VWR® VAPOUR-Line, Vertical Autoclaves with Exhaust Air Filtration (Option FA)
Supplier: VWR
VWR®
481-0782EA 20440 EUR
481-0782 481-0783 481-0795 481-0784 481-0796 481-0785 481-0797 481-0786 481-0798 481-0787
VWR® VAPOUR-Line, Vertical Autoclaves with Exhaust Air Filtration (Option FA)
Sterilisers Autoclaves (Steam Sterilisers)
These autoclaves are ideal for media, equipment and waste sterilisation. Designed for safety level laboratories, they are equipped with exhaust air filtration (option FA). This protects users and the environment against germs and genetically-modified material which is blown out of the autoclave into the laboratory during the de-aeration phase.
  • Easy to install, operate and service
  • With or without integrated fast cooling for liquids
  • Exhaust air filtration
  • Depth filter with metal housing does not distort
  • Integrated batch counter
  • Inline sterilisation of filter element during cycle
  • Filter element can easily be replaced
  • Sterile filter element 0,2µm with manufacturer certificate for 200 batches
Please contact your local Avantor sales office for other options.
Certifications: TÜV approved safety locking mechanism