VWR® VAPOUR-Line Lite II Autoclaves
Supplier: Avantor
VWR®
481-0846EA 2630 EUR
481-0846 481-0847
VWR® VAPOUR-Line Lite II Autoclaves
Sterilisers Autoclaves (Steam Sterilisers)
Autoclave for occasional use. AISI 304 stainless steel housing, easy to maintain and disinfect, aluminium inner chamber and ABS insulating lid with locking system.
  • Microprocessor-controlled automatic thermal regulation
  • Digital temperature and sterilisation time display
  • Automatic ventilation valve and electronic temperature cut-off safety protection system
  • Reinforced heating element and PT100 probe
  • Automatic sterilisation programs
  • Available with or without integrated printer
Instructions are GLP and EN 17205 compliant.
Ordering information: Basket is not supplied with the autoclave. Please order separately.