VWR® EML 200 Test Sieve Shakers
Supplier: Avantor
VWR®
510-2037EA 2760 EUR
HAVE209882094 HAVE209882193 HAVE209882216 HAVE209882117 510-2037 510-2039 510-2038 HAVE209882230 HAVE205323058 510-2051 HAVE209882056 510-2050 HAVE205323010 HAVE205323027 510-2046 510-2045 510-2048 510-2047 510-2049 HAVE209882209 HAVE205322990 HAVE209882186 510-2040 HAVE205323041 510-2042 HAVE209882223 510-2041 510-2044 510-2043 HAVE209882247
VWR® EML 200 Test Sieve Shakers
Shakers Sieve Shakers
The EML 200 series offers different types of units. All models are suitable for 50 to 203 mm Ø sieves, a maximum sample/batch load of 3 kg, and have a cast aluminium housing and improved clamping system.
  • Digital timer: 0 to 99 minutes or constant operation
  • All shakers have a cover with inspection glass which allows samples to be viewed during sieving
EML 200 Pure units are ideal for standard analysis:
Feature two given regulated amplitudes to select from: Fine or coarse
Operation in set intervals of 10 seconds
Classic 'eco' clamping system
EML 200 Premium units are suitable for users requiring precise and reproducible sieve analyses:
Extra quiet electromagnetic drive
Freely selectable amplitudes up to 3 mm
Extra large, easy to use display and simple logical operation
During sieving the remaining sieving time is shown on the display
Up to 49 different sieving programs can be stored in the memory
Option of Classic or HAVER TwinNut® quick release clamping system
EML 200 Premium Remote models also offer wet sieving, features as Premium units.
Integrated full cone nozzle
Separate controller which can be wall mounted
Ordering information: Acceptance test certificate 3.1 according to DIN EN 10204 and IQ/OQ documentation for are available as optional accessories for all models, for details please contact your local Avantor sales office.