Centrifugal filtration units, Vivaspin® 500
Supplier: Sartorius
Vivaspin®
512-3736EA 122 EUR
512-3736 512-4003 512-3740 512-2837 512-4001 512-4027 512-2838 512-3744 512-3743 512-4025 512-4005 512-3745 512-3739 512-4002 512-3737 512-4026 512-4004 512-3738
Centrifugal filtration units, Vivaspin® 500
Filters Centrifugal Filters
Vivaspin® centrifugal concentrators are disposable ultrafiltration devices for the concentration of biological samples. Vivaspin® 500 centrifugal filter units offer a simple, one step procedure for sample concentration with volumes from 5 to 500 µl. They can effectively be used in fixed angle rotors accepting 2,2 ml centrifuge tubes.
  • High retentate recovery >90%
  • Vertical membrane design for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
The patented vertical membrane design and thin channel filtration chamber (US 5,647,990), minimises membrane fouling and provides high speed concentrations.