Centrifugal filtration units, Vivaspin® 6
Supplier: Sartorius
Vivaspin®
512-4029EA 210 EUR
512-4029 512-3781 512-3778 512-3779 512-3775 512-4104 512-4103 512-3777 512-4017 512-4011 512-3776 512-4012 512-3782 512-4015 512-4014 512-4013
Centrifugal filtration units, Vivaspin® 6
Filters Centrifugal Filters
Vivaspin® 6 centrifugal concentrators have been developed to offer increased volume flexibility and performance. Vivaspin® 6 can process 6 ml in either swing bucket or fixed angle rotors accepting standard 15 ml conical bottom test tubes.
  • High retentate recovery >90%
  • Vertical membrane design for ultra-fast concentration
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
Vivaspin® 6 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. The remaining volume is easy to read off the printed scale on the side of the concentrator and the modified dead stop pocket further simplifies direct pipette recovery of the final concentrate.