Bottle top vacuum filtration systems
Supplier: Corning Life Sciences
513-3356EA 309.73 EUR
513-3356 513-3348 513-3352 734-1846 514-0169 734-1844 734-1861 734-1845 513-3357 513-3347 513-3358 514-0168 734-1860 513-3355 734-1821 513-3359 513-3354 513-3350 734-1900 513-3349 513-3346 734-1901
Bottle top vacuum filtration systems
Filters Bottle Top Filters
Vacuum filter systems available in four sizes: 150 ml; 250 ml; 500 ml and 1 L. Adapters are colour coded by membrane type for easy product identification: Polyethersulfone (PES) yellow; cellulose acetate (CA) orange; cellulose nitrate (CN) blue; nylon (NY) red.
  • Angled hose connector simplifies vacuum line attachment
  • Receiver bottles feature easy grip sides for improved handling
  • Individually packaged, sterile, certified non pyrogenic
  • Caps for receiver bottles are sterile and individually packaged
  • Extra plastic storage bottles are available
* 500 ml funnel with 70 mm membrane