Centrifugal devices, Microsep™ Advance
Supplier: Pall Laboratory
Microsep™
516-0376EA 216 EUR
516-0376 516-0374 516-0485 516-0373 516-0371 516-0370 516-0378 516-9920 516-0381 516-0382 516-0377 516-0375 516-0383 516-0372
Centrifugal devices, Microsep™ Advance
Filters Centrifugal Filters
These centrifugal devices provide a precise, quick recovery of microlitre volumes of concentrate.
  • High recoveries, typically greater than 90%
  • Deadstop to prevent samples from spinning to dryness
  • Fits centrifuges that accept standard 17×100 mm tubes
Applications: Concentrate dilute protein samples prior to electrophoresis. Exchange buffer and remove salt in samples. Remove proteins and particulate from samples for HPLC analysis of drugs, amino acids, and antibodies. Isolate low molecular weight compounds from fermentation broths for natural product screening. Recover biomolecules from cell culture supernatants or lysates. Clarify samples with gross particulate.