Centrifugal devices, Jumbosep™
Supplier: Pall Laboratory
Jumbosep™
516-8162EA 213 EUR
516-8162 516-8159 516-8160 516-0581 516-9342 516-8161
Centrifugal devices, Jumbosep™
Filters Centrifugal Filters
Convenient and reliable concentration, purification and diafiltration of 15 to 60 ml biological samples. Concentrate 60 ml sample volumes to 4 ml in 30 minutes.
  • High recoveries, typically greater than 90%
  • Built-in deadstop prevents spinning to dryness
Can be used with rotors for 250 ml tubes. Economical device as sample reservoir and filtrate receiver can be sanitised or autoclaved and reused.
* without membrane insert