Centrifuge tubes
Supplier: Greiner bio-one
391-3471EA 232 EUR
391-3471 391-3478 391-3479 391-3466 525-0412 525-0411 525-0410 391-3475
Centrifuge tubes
Tubes Centrifuge Tubes
In transparent PP or PS, with HDPE screw cap, sterile or non-sterile, round or conical bottom.
  • Non-autoclavable (except tubes 391-3478: autoclavable without cap)
  • Tubes 391-3478 and 391-3475 equipped with a screw cap requiring ¹/₃ turn to open
  • Free from DNase, RNase and human DNA, non pyrogenic and non cytotoxic
  • Cap colour: blue (except tubes 391-3471 and 391-3478: white, and tubes 391-3479: black)