Centrifuge tube filters, Costar® Spin-X®
Supplier: Corning Life Sciences
Costar® Spin-X®
525-3416EA 402.15 EUR
525-3416 612-5201 525-0018 525-0020 525-0017 525-0019 525-3415
Centrifuge tube filters, Costar® Spin-X®
Filters Centrifugal Filters
These centrifuge tube filters consist of a membrane containing filter unit within a microcentrifuge tube.
  • Removal of bacteria, cells and particles from liquids
  • HPLC sample preparation
  • DNA removal from agarose or acrylamide gels