VWR®, Snap Ring Vials ND11
Supplier: VWR
VWR®
548-0473AEA 65.1 EUR
548-0473A 548-0471A 548-0120A 548-0895A 548-0451A 548-0011A 548-8014A 548-0429A 548-8016A 548-2367A 548-1930A 548-2175A 548-2361A 548-1441A 548-1956A 548-1859A 548-0454A 548-0472A 548-0422A 548-3285A 548-0012A 548-2366A 548-8015A 548-2368A 548-1852A 548-2192A 548-1472A 548-2360A 548-1860A 548-0470A
VWR®, Snap Ring Vials ND11
Vials
Snap ring ND11 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polyethylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
  • Wide opening
Snap ring ND11 vials constructed from type I borosilicate glass with low metal content. Vials are available in amber or clear version with a flat bottom to ensure compatibility with inserts. Multiple cap colours mostly available with assembled septa are engineered to prevent sample contamination while providing consistent, secure seals. Furthermore, snap ring vials ND11 are available in amber and transparent plastic version.