Gas-tight syringes, 1 to 100 ml
Supplier: SGE (Trajan)
548-0949EA 73.8 EUR
548-0949 549-0334 549-0333 549-0344 SGEA008500CAL 549-0332 549-0343 548-0948 549-0342 549-0341 549-0340 SGEA008100CAL SGEA008962CAL 549-0339 549-0338 549-0337 549-0336 SGEA008102CAL 549-0335 SGEA008025CAL 548-0952 548-0950 548-0951
Gas-tight syringes, 1 to 100 ml
Syringes Syringes for Chromatography
Constructed from borosilicate glass, PTFE plunger tips and Kel-F® or PTFE Luer cones where applicable.
  • Assembled with an interference fit between plunger tip and barrel to effect the gas sealing properties of the syringe
  • Plungers are replaceable on these syringes
  • Suitable for manual and dispenser/diluter application
Barrel OD: 1 ml = 9 mm; 2,5 ml = 11 mm; 5 ml = 14 mm; 10 ml = 18 mm; 25 ml = 27 mm; 50 ml = 32,8 mm; 100 ml = 40,8 mm