VWR®, HPTLC Glass Plates, Silica, Unmodified Layers
Supplier: VWR
VWR®
551-0019EA 154 EUR
551-0019 551-0008 551-0026 551-0022
VWR®, HPTLC Glass Plates, Silica, Unmodified Layers
TLC Plates
These HPTLC glass plates use an optimised particle size which results in better separation efficiency.
  • Short migration distance
  • Increased detection sensitivity
  • Lower sample amount needed
Indicator: Manganese activated zinc silica with green fluorescence for short wave UV (254 nm).
Binder: Stable in almost all organic solvents.
Particle size: 2 to 10 µm