VWR®, TLC Glass Plates, Silica, Unmodified Layers
Supplier: VWR
VWR®
551-0006EA 157 EUR
551-0006 551-0017 551-0013 551-0024 551-0014 551-0031
VWR®, TLC Glass Plates, Silica, Unmodified Layers
TLC Plates
These TLC glass plates are suitable for easy and quick sample applications.
  • High sample throughput in short time
  • Pilot product for HPLC or Flash chromatography
  • Suitable for screening tests
Indicator: Manganese activated zinc silica with green fluorescence for short wave UV (254 nm).
Binder: Stable in almost all organic solvents.
Particle size: 5 to 17 µm