VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR
VWR®
551-0018EA 58.9 EUR
551-0018 551-0005 551-0020 551-0032 551-0021
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
TLC Plates
These TLC aluminium plates are suitable for easy and quick sample applications. They are flexible and can be cut to match different plate size requirements.
  • High sample throughput in short time
  • Pilot product for HPLC or Flash chromatography
  • Suitable for screening tests
Indicator: Manganese activated zinc silica with green fluorescence for short wave UV (254 nm).
Binder: Stable in almost all organic solvents.
Particle size: 5 to 17 µm