Precolumn filters, ColumnSaver
55215-UEA 254 EUR
55215-U 55214-U
Precolumn filters, ColumnSaver
Filters Precolumn Filters and Frits
Precolumn filter kit has a standard 10-32 fitting and fits direct to most column types without introducing an additional dead volume.