Weighing bottles, glass
Supplier: Lenz
611-9324EA 138.36 EUR
611-9324 611-1239 611-9323 611-1238 611-1237 611-9321 611-1236 611-9325 611-9319 611-9308 611-9318 611-9320
Weighing bottles, glass
Bottles Weighing Bottles
DURAN®, borosilicate glass.
  • With interchangeable cap
  • Exterior joint improves material transfers and reduces risk of material contamination of cap
  • Available in low, medium or tall forms