VWR®, Safety Pipette Fillers, Standard/Universal/Flip
Supplier: VWR
VWR®
612-1947EA 21.8 EUR
VWRU53497-055 612-1947 612-1931 612-1920 612-2008 612-2004 612-1930
VWR®, Safety Pipette Fillers, Standard/Universal/Flip
Pipettes Pipette Bulbs
Natural rubber.
  • For measuring and for volumetric pipettes up to 10 ml (standard) or 100 ml (universal/flip)
  • Standard/universal: Three ventilation valves
  • Flip: Only two operating points for easier usage (release is through an automatic removable valve)