Pipette tips for Eppendorf®
Supplier: Greiner bio-one
613-0389EA 624 EUR
613-0389 GREI765280_960
Pipette tips for Eppendorf®
Pipette Tips
These pipette tips feature a shorter tip collar and are recommended for use with Eppendorf® pipettors.
  • Bevelled edge increases pipetting accuracy
  • Manufactured from high-grade PP to give the optimum in performance and fit
  • Free of heavy metals
  • Autoclavable
Ordering information: Matching racks for bulkware tips can be ordered separately.