Single-use syringes, 3-piece, HENKE-JECT®
Supplier: Henke-Sass, Wolf
HENKE-JECT®
615-0041EA 20 EUR
615-0041 615-0042 613-2051 613-2050 HSWA5100.090D0 613-6914 613-6913 613-3864 613-2053 613-2052 613-6079 613-2794 613-1586 613-2040 HSWA5050.X90D0 HSWA5202D000V0 613-6080 613-2049 613-2042 613-2041 613-2044 613-2043 613-2046 613-2045 613-3135 613-2047
Single-use syringes, 3-piece, HENKE-JECT®
Syringes Syringes, Medical
Disposable, three-piece syringes with PP barrel and piston, polyisoprene stopper, latex-free.
  • Lubricated synthetic rubber piston allows a smooth draw
  • Available in non sterile version (bulk) upon request
Without needles: Luer lock types are compatible for use with most common syringe pumps. Please ask for further information.
With fixed needles: Needle tube permanently attached in syringe, no dead space, safety stop, suitable for dosage of very small volumes.
Packaging: Individual sterile blisters.