Dial thermometers
Supplier: VWR, part of Avantor
VWR®
620-0867EA 17 EUR
620-0867 620-0868 620-0871 620-0874
Dial thermometers
Thermometers Analogue Thermometers
These thermometers are adapted for applications in heating, ventilation, air conditioning, food and beverage processing and storage, pharmaceutical and educational use.
  • Quick and accurate
  • Easy to read instrument
  • Suitable alternative to mercury thermometers
Immersion depth: Minimum immersion is 50 mm (2") in liquids or 100 mm (4") in gases
Stainless steel probe
Delivery information: 125 mm stem thermometers have a protective probe cover with pocket clip. 200 mm stem thermometers have a beaker clip.