VWR® Traceable® Large Digit, Triple-Display Thermometers

Supplier: VWR


VWR®
620-1584EA 116 EUR
620-1584 620-1585 VWRU89094-770
VWR® Traceable® Large Digit, Triple-Display Thermometers
Thermometers Digital Thermometers
These jumbo size display thermometers are suitable for monitoring freezers, water baths, heating blocks, incubators and refrigerators. A bottle insulates the sensor from transient temperature changes when the refrigerator door is opened. Solution is GRAS (Generally Recognised As Safe) by the FDA (Food and Drug Administration).

  • Triple display: Maximum/minimum and current temperatures
  • Out of range alarm programmable in 1,0 °C increments

Bottle probe in non toxic glycol solution or waterproof bullet probe (20 mm).

Bottle probe sensor: Temperature-buffered glycol bottle probe mimics solution. Bottle and solution insulate sensor from rapid changes, as an example, when a refrigerator door is opened. Solution mimics temperature of stored samples. Probe is sealed in a miniature bottle filled with nontoxic glycol. Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration).

Bullet probe sensor: Small sized plastic probe designed for use in air and liquids, the sensor and cable may be completely immersed.

Certifications: Traceability to NIST. Fulfils CDC and vaccine thermometer requirements. Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.

Delivery information: Supplied with Traceable® certificate of calibration, slot for wall mounting, VELCRO® and magnetic strips to attach to any surface, AAA battery and 3 metre cable.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR