VWR® Traceable®, Thermometers for Refrigerators and Freezers
Supplier: VWR
VWR® Traceable®
620-2211EA 253 EUR
620-2211 VWRU89094-750 620-2213 620-2212 VWRU89369-152 620-2215 620-2214 620-2217 620-2216
VWR® Traceable®, Thermometers for Refrigerators and Freezers
Thermometers Digital Thermometers
These digital thermometers are designed for monitoring temperatures in refrigerators, freezers, water baths, heating blocks and incubators. Bottle probes are used in monitoring storage conditions for vaccines or other important samples.
  • Fixed 1 minute logging interval
  • Stores up to 10 unique alarm events
  • Capable of storing over one million observations
  • Raw data on unit cannot be overwritten or modified
  • Status: Low battery, low memory, USB data transfer and active alarm state
Bottle probe in non toxic glycol/bullet sensor/stainless steel probe with 3 metre cable.
Certifications: Traceable to NIST. Calibration compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001. Meets current CDC requirements and additional recommendations for vaccine storage and monitoring. Can be used in environments requiring 21 CFR 11 compliance.
Delivery information: Supplied with batteries, universal adapter, stand, VELCRO®, magnetic strips, wall mount, Traceable® certificate of calibration and batteries.