Binocular microscopes, STB series
630-0768EA 0 EUR
VWRI630-0768
Binocular microscopes, STB series
These STB series of stereo microscopes can be used in education, life science applications, and electronics inspection.
  • Provides a large, three dimensional image of the specimen
  • Working distance 88 mm
  • Adjustable interpupillary distance
  • Slip-clutch mechanism prevents gear damage