VWR® VisiScope® 250, Stereo Microscopes
Supplier: VWR
VWR® VisiScope®
630-1577EA 506 EUR
630-1577
VWR® VisiScope® 250, Stereo Microscopes
Microscopes
Reliable microscopes for educational purposes. Particularly recommended for secondary and higher schools. SZB250 and SZT250 have a 0,7× to 4,5× zoom range (6,43:1 zoom ratio), which is ideal for routine inspections.
  • Compact and slim units thanks to V-shaped Greenough optical system
  • Incident and transmitted light with two efficient LEDs
  • Efficient LED energy saving lamp
  • Adjustable angle of the incident illuminator
  • Integrated handle for easy lifting and carrying of the microscope
Ordering information: Supplied ready to use with eyepieces, objective and multi-plug power supply.