Upright microscopes, VisiScope® 800, VWR®
Supplier: VWR, part of Avantor
VWR® VisiScope®
630-1969EA 156 EUR
630-1969 630-1968 630-1967 630-1941 630-1940
Upright microscopes, VisiScope® 800, VWR®
Microscopes
VisiScope 800 laboratory microscopes are designed for research projects.
  • Die-cast frame with high stability and ergonomics
  • Transmitted light observations
  • High-efficiency pure white LED
  • IOS objectives with anti-fungus treatment
Ordering information: A full range of accessories are available. Please contact your local Avantor sales office for further details.
Delivery information: Supplied ready to use with eyepieces, objectives, dust cover and power supply.