Upright microscopes, VisiScope® ML134 R LED / BL134 R LED
Supplier: VWR, part of Avantor
VisiScope®
630-2981EA 566 EUR
630-2981 630-2980
Upright microscopes, VisiScope® ML134 R LED / BL134 R LED
Microscopes
These monocular and binocular microscopes are ideal for educational purposes.
  • Energy efficient LED illumination with light intensity control for constant pure-white colour temperature
  • Coaxial coarse and fine focusing mechanism with limit stop to prevent contact between objective and specimen
  • Adjustable tension of coarse focusing knob
  • Smooth movement of the mechanical stage enables precise, accurate positioning of the slide
  • N-plan objectives with anti-fungus treatment
  • Cordless operation thanks to rechargeable Li-Ion battery (charging time: 6 hours)
Delivery information: Supplied ready to use with eyepieces, objectives and external power supply with EU, UK and US plug for charging the Li-Ion battery.