Stereo microscopes, VisiScope® 260
Supplier: VWR, part of Avantor
VWR®
630-3074EA 386.4 EUR
630-3074 630-3073 630-3072
Stereo microscopes, VisiScope® 260
Microscopes
The cordless VisiScope 260 series of microscopes include binocular stereo microscope, binocular stereo zoom and trinocular stereo zoom versions. Ideal for higher education and laboratory applications.
  • Brightfield observation method for transmitted and incident light
  • Greenough optical system
  • 360° rotating head
  • Long lasting EcoLED illumination
  • Uses Ni-MH rechargeable batteries for up to 12 hours of working time
The stereo zoom models feature a parfocal achromatic zoom.
Ordering information: Supplied ready to use with eyepieces, objective, multi-plug power supply, dust cover and user manual, digital version (downloadable).