Upright microscopes, VisiScope® 200
Supplier: VWR, part of Avantor
VWR® VisiScope®
630-3295EA 311.2 EUR
630-3295 630-3294 630-3293 630-3292 630-3291
Upright microscopes, VisiScope® 200
Microscopes
Reliable, advanced microscopes for educational purposes. Particularly recommended for secondary schools.
  • Powered illumination system with adjustable intensity – efficient LED energy saving lamp
  • Constant pure-white colour temperature at all intensity levels
  • Large opening behind the nosepiece for use as a handle
  • Optical components treated to prevent the formation of moulds and fungi
Certifications: Objectives conform to DIN standards. Strict controls on materials and production processes optimised by ISO 9001:2008 quality system.
Delivery information: Supplied ready to use with eyepieces, objectives and multi-plug external power supply.