Microscope solution for materials analysis, DM6 M LIBS
Supplier: Leica
Leica®
630-3397EA 0 EUR
630-3397
Microscope solution for materials analysis, DM6 M LIBS
Microscopes
See the particles and know their composition at the same time. 2-in-1 materials analysis solution, combining optical microscopy and chemical analysis by laser induced breakdown spectroscopy (LIBS), leads to an overall more efficient and cost-effective technical cleanliness analysis.
  • Cleanliness analysis with a 2-methods-in-1 solution - LIBS
  • Save 90% time for chemical analysis compared to typical methods involving electron microscopy
  • Quick, confident decision making based on reliable information about the material, both visual and chemical
Cleanliness analysis is instrumental in assuring product quality and reliability in a number of industries, such as automotive and transportation. A 2-in-1 solution can significantly reduce the cost and time for a cleanliness analysis workflow, as particle image and composition data are obtained at the same time. The obtainment of both visual and chemical data simultaneously enables easier identification of the sources of contamination.
The complete analysis workflow is faster in comparison to other methods, such as scanning electron microscopy (SEM), because there is no sample preparation; no sample transfer to another device is required; and the filter sample is always analyzed in air under atmospheric conditions.
Both visual and chemical data for analyzed particles are readily available when using a 2-in-1 solution which leads to confident decision-making in less time concerning further actions to be taken during production, quality control, and failure analysis.
Ordering information: For more information on LIBS, please contact local Avantor representative.