LED illumination system, VisiLED
Supplier: SCHOTT Fibre Optics
VisiLED
630-3500EA 670 EUR
630-3500 630-3181
LED illumination system, VisiLED
Lights Cold Light Sources
Professional illumination for stereo microscopy. The VisiLED product line offers modular, homogeneous, and shadow-free LED illumination for stereo microscopy and is not just an accessory but a professional illumination system for even specialised tasks up to the highest magnifications.
  • Flexible options for strong contrast
  • VisiLED product line offers powerful light heads for tasks up to the highest magnification
  • Darkfield options and control of individual segments enable a number of contrasting methods to make the invisible visible
  • Powerful luminance
  • Intense light output for specialized tasks
  • Intelligent light heads
  • Actively controlled LED temperature
  • Maintenance-free
Moulding the lighting setup to each application, a number of high performance ring lights and light stages enable high intensity illumination, or even a combination of techniques. The system offers controllable light heads in segments for contrasting methods, as well as easy combination of brightfield and darkfield illumination, or of incident light with transmitted light. Illumination parameters can be stored in memory positions, for reproducible mixed light conditions.
MC1500 - Multifunction control with memory simultaneously for two light heads, plus strobing, external triggering or flashing of the LED light heads.
MC1000 - Standard control, compact unit allows settings of various illumination parameters including light intensity, Continuous dimming via potentiometer; five preset segment modes and change of illumination direction. Easy to use.
MC750 - Basic control for low contrasting requirements. Continuous dimming via potentiometer and over-temperature protection of the LED light head.
Ring lights: The VisiLED brightfield ring lights offer homogeneous and shadow-free illumination in a robust metal housing and well designed heat sink to enable a maximum brightness of 200 kLux. The VisiLED brightfield ring lights S40-75, S80-55, S80-25 and S80-65 N offer different brightness levels, working distances and mounting diameters for various stereo microscopy objectives. In combination with the available accessories, like diffusers, polariser sets and adaptor rings, different illumination modes are possible. The VisiLED slim ring light enables a maximum brightness of 65 kLux. The incredibly thin and light design with an outer diameter of just 82 mm is optimal for unconfined object preparation and provides ample space for the usage with objective revolvers. The VisiLED darkfield ring light S40-10 D enables a maximum brightness of 130 kLux and offers the possibility of real incident darkfield illumination. In addition, it can be combined with all brightfield ring lights using the brightfield-darkfield adapter kit.
Transmitted light stage : Professional incident and darkfield illumination. The VisiLED backlight offers homogeneous illumination with a maximum light density up to 20 000 cd/m. The VisiLED darkfield backlight TLS-DF offers the possibility of transmitted darkfield illumination.
Ordering information: Power supply and power cord need to be ordered separately.