Staining jars
Supplier: Kartell
631-9131EA 128 EUR
631-9131
Staining jars
Jars Staining Jars
Made from PMP (TPX).
This rectangular jar is supplied with a lid and can hold 10 slides or 20 slides back-to-back.