Disposable cuvettes
Supplier: Kartell
634-0326EA 16.1 EUR
634-0326 634-8535 634-8537 634-8536 634-8531 634-8530 634-8533 634-8532
Disposable cuvettes
Cuvettes Spectrophotometer Cuvettes
Disposable cuvettes made from optical polystyrene (PS) or PMMA with a transmission value at 300 nm of 70%.