VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
Supplier: VWR
VWR®
662-2270EA 423 EUR
662-2270 662-1350 662-2269 662-2268
VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
Electrochemical Meters pH and Multi-Parameter Meters
These economical, robust and waterproof pH meters are supplied in complete, ready to use kits that incorporate a robust carry case, electrode and temperature sensor with different cable lengths and buffers.
Warranty: 3 years for instruments and 6 months for electrodes/probes/cables
  • IP 67 waterproof housing, large display and easy to use keypad
  • Automatic/manual temperature compensation
  • 50 or 250 data sets reviewable memory
  • 1000 hour battery life; auto-shut off function (30 minutes of non use)
  • Electrode offset recognition; auto-lock and hold of readings on display
Additional functions of pH 110 M:
- Extended memory capacity (250 data sets)
- Waterproof USB port for transferring data to a PC (no software required)
- Real time clock for saved data
- Recalibration reminder (selectable from 0 to 60 days)
Designed for quick, accurate measurements in various applications in industries, laboratories or in the field.
Warranty: 3 years for instruments and 6 months for electrodes/probes/cables
Ordering information: Supplied in a carry case with 1 m or 4 m cable, pH/°C sensor and calibration buffers (4,01/7,00/10,00).