VWR® PerfectBlue, Semi-dry electroblotters
Supplier: VWR
VWR®
700-1220EA 1700 EUR
700-1220 700-1219
VWR® PerfectBlue, Semi-dry electroblotters
Blotting Apparatus
PerfectBlue semi-dry electroblotters are designed for clean, rapid and effective transfer of proteins and nucleic acids from polyacrylamide or agarose gels to membranes. Suitable for use with Western, Southern and Northern blotting, these semi-dry blotters reduce many lengthy procedures down to around an hour.
Blotting chamber
Highly durable Plexiglass® construction
Totally removable cover which attaches flat to reduce overall unit storage size
Oversized, easy to handle screws for fumble-free and safe cover adjustment
High quality gold plated plug contacts for optimal electrical contact.
Electrodes
Solid electrode plates for an even transfer over the entire gel surface
High grade stainless steel cathode and platinum coated titanium anode for maximum transfer efficiencies
Sturdy cover for enhanced user safety
Design of lid prevents accidental wrong orientation of electrical field.