Cell culture storage bottles, polystyrene, Corning®
Supplier: Corning
Corning®
734-1897EA 107.28 EUR
734-1897 700-1783 734-1847 734-1825 734-1824 700-1785 700-1787 700-1786
Cell culture storage bottles, polystyrene, Corning®
Bottles Cell Culture Bottles
Disposable polystyrene bottles for storage of media, buffers and other aqueous solutions.
  • Low profile, easy grip style has sides that facilitate handling
  • Plug seal caps (45 mm) provide an airtight seal and help minimise the risk of contamination
  • Bottles can be used with Corning® vacuum filter systems
  • Sterile, certified non-pyrogenic