Swabs with transport medium for viruses and Chlamydia
Supplier: COPAN Diagnostics
710-0454EA 48.6 EUR
710-0454 710-0453 710-0451 710-0450
Swabs with transport medium for viruses and Chlamydia
Swabs Transport Swabs
Liquid transport medium is contained in a sponge carrier located in the bottom of the tube.
  • Special colours of caps to make differentiation easy
  • Simple insert swab into the tube after collecting sample
  • Swab tip placed in direct contact with the medium soaked sponge
Certifications: ISO 13485. ISO 9001:2008.
Packaging: Individually sealed in a peel-pouch. 25 swabs per box, 10x25 swabs per case.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904