Count plates, Petrifilm™
Supplier: 3M
710-0779EA 54.5 EUR
XXXX6406 XXXX6416 XXXX06457-MEM XXXX06404 XXXX06402 710-0779 710-3408 710-3409 710-0849 710-3406 XXXX06493 XXXX06450 710-3407 710-0778 710-0777 XXXX06434 XXXX6400 710-0780 XXXX06490
Count plates, Petrifilm™
Media for Microbiology Prepared Media for Microbiology
3M™ Petrifilm™ is an extremely cost-effective, practical and reliable method for testing equipment, raw materials, food products and their manufacturing environment; it also enables more frequent monitoring of critical control points. Fast and easy-to-approve results provide you with the information required to make the right decisions. The end result is an increase in product quality, improved process control and an overall reduction in operating costs.
  • Fast and accurate results in 48 hours
  • Space-saving
  • 3M™ Petrifilm™ plates can also be read in four seconds using the 3M™ Petrifilm™ plate reader