Mini tube systems (MTS) caps, Axygen®
Supplier: Corning
Axygen®
731-0224EA 166 EUR
731-0224 731-0221 731-0222 731-0223
Mini tube systems (MTS) caps, Axygen®
Closures Test/Sample Tube Closures
Axygen® mini tube systems are available in 8-cap or 12-cap strip format.
  • Molded-in condensation rings on the lid minimise cross contamination