VWR®, Strips of PCR Tubes and Caps
Supplier: VWR
VWR®
732-1517EA 76.5 EUR
732-1517 VWRU76402-854 732-1519 732-1518 731-0367 732-0550 732-3484 732-3485 732-0552 732-1520 732-0554 732-0553 732-1521 732-3608 732-1506 732-3609 VWRU93001-118 VWRU20170-000 VWRU20170-004 732-3391 211-0381 VWRU20170-002 211-0338 732-3499 732-3610 VWRU82006-606 VWRU93001-118_U 732-0545 211-0339 VWRU76318-796 732-0546
VWR®, Strips of PCR Tubes and Caps
Tubes PCR Tubes
These PCR tube strips and caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
  • Certified free from DNase, RNase and human DNA
  • Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.
Colour: Natural