Strips of PCR tubes and caps
Supplier: VWR, part of Avantor
VWR®
732-0554EA 141 EUR
VWRU20170-004 732-0554 732-3485 211-0339 VWRU76318-796 211-0338 732-3608 732-3610 732-1506 VWRU82006-606 VWRU20170-000 VWRU93001-118_U 732-0553 732-0552 732-1517 732-1518 VWRU93001-118 VWRU20170-002 732-3499 732-1521 732-0546 732-0550 732-3484 732-1519 732-3609 732-0545 VWRU76402-854 732-3391 211-0381 732-1520 731-0367
Strips of PCR tubes and caps
Tubes PCR Tubes
These PCR tube strips and caps are made of PP resin and are designed to fit most popular brands of thermal cyclers.
  • Certified free from DNase, RNase and human DNA
  • Manufactured from polypropylene under strict quality control guidelines
Available with domed or flat caps.
Colour: Natural