Filter plates, 96-well, Corning® FiltrEX™
Supplier: Corning
Corning® FiltrEX™
732-5509EA 1663.37 EUR
732-5509 732-5511 732-5510 732-5513 732-5512
Filter plates, 96-well, Corning® FiltrEX™
Microplates Filter Plates
FiltrEX™ filter plates meet the industry standards for plate dimensions. The rigid side walls make the plate ideal for automation and the wide skirt accepts barcodes. Individual filter discs are encapsulated in the plate by a process that ensures 100% integrity of each well. The design of the nozzle prevents sample wicking and cross-contamination. Glass fibre filter plates can be used for a variety of applications, such as plasmid isolation, DNA purification, PCR clean-up or receptor/ligand binding assays. The low-binding hydrophilic PVDF membrane can be used for lysate clarification, protein kinase assays, or bead- or resin-based separation assays.