Clear bottom black and white polystyrene microplates, 96 well, Corning®
Supplier: Corning
Corning® NBS™ Corning® CellBIND® Corning® Special Optics Corning®
734-4160EA 1206.59 EUR
734-4160 734-4172 734-4173 734-2293 734-1609 734-1608 CORN3904BC 734-1612 734-1610 734-1613 734-4069 734-4157 734-4158 734-4159 732-5523 732-5519 734-4154 732-5518 734-4155 734-1661 734-1660 732-5516 734-4156
Clear bottom black and white polystyrene microplates, 96 well, Corning®
Microplates Immunology Microplates and Strips
PS, flat bottom.
  • Bottoms are 60% thinner than conventional PS plates, resulting in lower background fluorescence and enabling readings down to 340 nm
  • Opaque walls prevent well-to-well crosstalk
  • Optically clear flat bottom profile permits direct microscope viewing
NBS™ = Non Binding Surface.