Taq DNA polymerase master mix
Supplier: VWR Life Science
733-1317EA 840 EUR
733-1317 733-2542 733-2543 733-2540 733-1316 733-1314 733-1315
Taq DNA polymerase master mix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VWR® Taq DNA Polymerase Master Mix is a ready to use 1,1X or 2X reaction mix. Simply add primers, template and water to carry out primer extensions and other molecular biology applications.
Tests = Reactions