VWR®, Red Taq DNA polymerase master mix
Supplier: Avantor
VWR®
733-1320EA 826 EUR
733-1320 733-1321 733-2132 733-2131 733-2130 733-1319 733-1318 733-2547 733-2546 733-2545 733-2544
VWR®, Red Taq DNA polymerase master mix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VWR® Red Taq DNA Polymerase Master Mix, which also contains an inert red dye, can be directly loaded onto an agarose gel without addition of electrophoresis loading buffers.
Tests = Reactions