Red Taq DNA polymerase
Supplier: VWR Life Science
733-2408EA 94.5 EUR
733-2408 733-1834 733-2409 733-1323
Red Taq DNA polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Red Taq DNA Polymerase is a blend of Taq DNA polymerase combined with an inert red dye. The dye enables quick visual recognition of reactions to which enzyme has been added, as well as confirmation of complete mixing.
  • Ideal choice for routine applications
  • High performance, thermostable DNA polymerase
  • Optimal for TA cloning
Taq DNA polymerase concentration: 5 Units/μl
10X Key Buffer: Tris-HCl pH 8,5; (NH₄)₂SO₂, 15 mM MgCl₂, 1% Tween® 20
EU = Units
Delivery information: VWR® Taq DNA Polymerase is usually supplied with Key Buffer. Key Buffer (NH⁴⁺) gives a superior amplification signal (high yield) minimising the need for optimisation of the Mg²⁺ concentration, or the annealing temperature in most primer-template systems.