Red Taq DNA polymerase master mix
Supplier: VWR Life Science
733-2544EA 229 EUR
733-2544 733-1319 733-2545 733-2131 733-2132 733-1318 733-1321 733-2547 733-1320 733-2546 733-2130
Red Taq DNA polymerase master mix
Nucleic Acid Reagents End-point PCR Enzymes and Kits
VWR® Red Taq DNA Polymerase Master Mix, which also contains an inert red dye, can be directly loaded onto an agarose gel without addition of electrophoresis loading buffers.
Tests = Reactions