VWR®, DNA loading buffers
Supplier: Avantor
VWR®
733-2575EA 42.5 EUR
733-2575 733-2574 733-2576
VWR®, DNA loading buffers
Stains Electrophoresis Stains
These DNA loading dyes are used to load DNA samples to agarose or SDS DNA gels for gel electrophoresis.
Suitable for TAE, TBE, SB and LB electrophoresis buffers.
Certifications: EC Regulation No. 1907/2006 (REACH).
Packaging: Each kit contains 5×1 ml.
Caution: Store at –20 °C for in vitro use only.