VWR®, Taq DNA polymerase, glycerol-free

Supplier: Avantor
VWR®
733-2875EA 0 EUR
733-2875 733-2874 733-2873
VWR®, Taq DNA polymerase, glycerol-free
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Highly concentrated Taq DNA polymerase glycerol free 50 U/µl is ideal for industrial use and manufactures of diagnostic PCR kits.

  • Suitable for automated high throughput testing
  • Suitable for applications involving freeze drying
  • Suitable for robot-aided pipetting
  • Ideal for kit manufacturers
  • Concentration: 50 U/µl
  • Fidelity: 1×Taq
  • Amplicon size: <5 kb

Taq DNA Polymerase Glycerol Free 50 U is a thermostable recombinant DNA polymerase, which exhibits very high activity in primer extension and other molecular biology applications. The enzyme is isolated from Thermus aquaticus and has a molecular weight of approximately 94 kDa.

Taq DNA Polymerase Glycerol Free has both a 5'→3' DNA polymerase and a 5'→3' exonuclease activity. The enzyme lacks a 3'→5' exonuclease activity (no proofreading ability). Taq DNA Polymerase Glycerol Free leaves an A' overhang, which makes the enzyme ideal for TA cloning.

The glycerol free formulation makes it especially well-suited for freeze drying and automatisation.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR