PS, sterile, with lid. Designed for cell culture, these dishes have a uniform surface chemistry, created by vacuum gas plasma treatment, which promotes cell attachment. The hydrophilic tissue culture surface contains a variety of negatively charged functional groups that support cell attachment and spreading.
- Flat and optically clear for distortion-free microscopic visualisation of cells
- Lids designed for optimal gas exchange
- Stacking rings allow for easier stacking and handling
- Cell performance tested to ensure consistent results
The design and frosted rim of the Easy-Grip dish improves handling of small dishes. The ability to pick up a small dish conveniently, without accidentally removing the lid, allows faster working and improves aseptic manipulation.